Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269738.3 | NP_001256667.1 | 8 - 9 | 1764 | 73 | |
NM_074449.6 | NP_506850.1 | 8 - 9 | 1989 | 73 | ||
NM_074450.6 | NP_506851.1 | 9 - 10 | 1937 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |