Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269741.1 | NP_001256670.1 | 5 - 6 | 652 | 73 | |
NM_001269742.2 | NP_001256671.1 | 11 - 12 | 1651 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |