Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxa-95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074470.4 | NP_506871.3 | 1 - 2 | 115 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |