Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sru-42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074493.3 | NP_506894.2 | 3 - 4 | 2260 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |