Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cgt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074570.5 | NP_506971.2 | 5 - 6 | 1062 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |