Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srj-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074645.1 | NP_507046.1 | 4 - 5 | 744 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |