Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxa-88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074664.5 | NP_507065.2 | 2 - 3 | 102 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |