Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp-33D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074675.4 | NP_507076.1 | 5 - 6 | 1238 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |