Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074719.5 | NP_507120.3 | 10 - 11 | 2119 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |