Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srr-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074779.5 | NP_507180.1 | 7 - 8 | 992 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |