Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srd-68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074870.2 | NP_507271.2 | 3 - 4 | 432 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |