Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322605.2 | NP_001309463.1 | 3 - 4 | 373 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |