Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpy-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029095.4 | NP_001024266.1 | 10 - 11 | 4901 | 64 | |
NR_002407.1 | 9 - 10 | 1324 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |