Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269843.3 | NP_001256772.1 | 4 - 5 | 384 | 63 | |
NM_001269844.2 | NP_001256773.1 | 4 - 5 | 384 | 63 | ||
NM_001269845.1 | NP_001256774.1 | 4 - 5 | 374 | 63 | ||
NM_001269846.1 | NP_001256775.1 | 4 - 5 | 374 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |