Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: par-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380857.1 | NP_001366977.1 | 1 - 2 | 159 | 94 | |
NM_001380858.1 | NP_001367285.1 | 1 - 2 | 148 | 94 | ||
GenBank mRNA | AF160189.1 | - | 1 - 2 | 182 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |