Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srg-34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075230.3 | NP_507631.1 | 4 - 5 | 638 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |