Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp-33D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075278.4 | NP_507679.2 | 5 - 6 | 1234 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |