Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: emb-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269902.2 | NP_001256831.1 | 3 - 4 | 718 | 114 | |
NM_001313364.2 | NP_001300293.1 | 3 - 4 | 718 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Caenorhabditis elegans Probe spans exons |