Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373193.1 | NP_001360177.1 | 4 - 5 | 336 | 63 | |
NM_075333.8 | NP_507734.4 | 8 - 9 | 1130 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |