Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gly-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029045.5 | NP_001024216.1 | 12 - 13 | 2268 | 73 | |
NM_075449.9 | NP_507850.2 | 8 - 9 | 1566 | 73 | ||
GenBank mRNA | AF031834.1 | - | 9 - 10 | 1820 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Caenorhabditis elegans Probe spans exons |