Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075468.6 | NP_507869.1 | 7 - 8 | 2329 | 92 | |
GenBank mRNA | Z38016.1 | - | 2351 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |