Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fre-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307796.3 | NP_001294725.1 | 3 - 4 | 599 | 74 | |
NM_075474.6 | NP_507875.1 | 3 - 4 | 597 | 74 | ||
GenBank mRNA | AY079165.1 | - | 3 - 4 | 587 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |