Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ssu-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075479.2 | NP_507880.1 | 1 - 2 | 253 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |