Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srz-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075490.2 | NP_507891.2 | 1 - 2 | 155 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |