Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fog-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047721.3 | NP_001041186.1 | 3 - 4 | 791 | 151 | |
NM_001047722.2 | NP_001041187.1 | 3 - 4 | 791 | 151 |
Gene Symbol: fbxa-114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075496.4 | NP_507897.1 | 3 - 4 | 774 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |