Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnj-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270018.2 | NP_001256947.1 | 6 - 7 | 694 | 129 | |
NM_001373209.3 | NP_001360578.1 | 6 - 7 | 701 | 129 | ||
GenBank mRNA | AF293972.1 | - | 6 - 7 | 714 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |