Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270021.4 | NP_001256950.1 | 15 - 16 | 2580 | 85 | |
NM_001270022.3 | NP_001256951.1 | 15 - 16 | 2542 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |