Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hif-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028720.5 | NP_001023891.2 | 5 - 6 | 1335 | 66 | |
NM_001028721.5 | NP_001023892.1 | 2 - 3 | 740 | 66 | ||
NM_001028723.4 | NP_001023894.2 | 4 - 5 | 1216 | 66 | ||
NM_001313065.3 | NP_001299994.1 | 4 - 5 | 1216 | 66 | ||
NM_075607.7 | NP_508008.4 | 5 - 6 | 1329 | 66 | ||
GenBank mRNA | AF364604.1 | - | 5 - 6 | 1344 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |