Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hif-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028721.5 | NP_001023892.1 | 3 - 4 | 1070 | 66 | |
NM_001028723.4 | NP_001023894.2 | 5 - 6 | 1546 | 66 | ||
NM_001373210.2 | NP_001359648.1 | 8 | 1868 | 66 | ||
NM_075607.7 | NP_508008.4 | 6 - 7 | 1659 | 66 | ||
GenBank mRNA | AF364604.1 | - | 6 - 7 | 1674 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |