Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: num-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028926.5 | NP_001024097.1 | 3 - 4 | 847 | 58 | |
NM_001028927.3 | NP_001024098.1 | 4 - 5 | 864 | 58 | ||
NM_001136432.5 | NP_001129904.1 | 6 - 7 | 1820 | 58 | ||
NM_001270029.3 | NP_001256958.1 | 3 - 4 | 822 | 58 | ||
NM_075620.6 | NP_508021.1 | 4 - 5 | 996 | 58 | ||
GenBank mRNA | AF286205.1 | - | 4 - 5 | 996 | 58 | |
AF286206.1 | - | 4 - 5 | 864 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |