Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dve-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029813.6 | NP_001024984.1 | 4 - 5 | 533 | 95 | |
NM_001351875.4 | NP_001338861.1 | 3 | 419 | 95 | ||
NM_001351876.3 | NP_001338860.1 | 3 - 4 | 427 | 95 | ||
NM_001383518.1 | 4 - 5 | 534 | 95 | |||
NR_147826.1 | 2 - 3 | 1113 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |