Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: che-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075705.6 | NP_508106.1 | 6 - 7 | 892 | 102 | |
GenBank mRNA | AJ011523.1 | - | 6 - 7 | 914 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |