Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001361781.3 | NP_001348781.1 | 2 - 3 | 415 | 84 | |
NM_075711.7 | NP_508112.1 | 2 - 3 | 415 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |