Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nas-38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373215.1 | NP_001359993.1 | 16 - 17 | 2099 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |