Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nas-38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373215.1 | NP_001359993.1 | 3 - 4 | 395 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |