Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: irk-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270047.4 | NP_001256976.2 | 10 - 11 | 931 | 87 | |
NM_001270048.3 | NP_001256977.1 | 10 - 11 | 939 | 87 | ||
NM_001307921.4 | NP_001294850.1 | 10 - 11 | 941 | 87 | ||
NM_001330888.3 | NP_001317907.1 | 11 - 12 | 1030 | 87 | ||
NM_001380895.1 | NP_001368610.1 | 11 - 12 | 1030 | 87 | ||
NM_001380896.1 | NP_001368348.1 | 11 - 12 | 1050 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |