Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: irk-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270047.4 | NP_001256976.2 | 9 - 10 | 843 | 80 | |
NM_001270048.3 | NP_001256977.1 | 9 - 10 | 851 | 80 | ||
NM_001307921.4 | NP_001294850.1 | 9 - 10 | 853 | 80 | ||
NM_001330888.3 | NP_001317907.1 | 10 - 11 | 942 | 80 | ||
NM_001380895.1 | NP_001368610.1 | 10 - 11 | 942 | 80 | ||
NM_001380896.1 | NP_001368348.1 | 10 - 11 | 962 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |