Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trap-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075749.8 | NP_508150.1 | 2 - 3 | 187 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |