Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nas-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075753.8 | NP_508154.2 | 2 - 3 | 289 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |