Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029433.7 | NP_001024604.1 | 10 - 11 | 1914 | 142 | |
NM_001029434.5 | NP_001024605.1 | 8 - 9 | 1668 | 142 | ||
NM_001313412.4 | NP_001300341.1 | 10 - 11 | 1960 | 142 | ||
NM_001313413.4 | NP_001300342.1 | 10 - 11 | 1907 | 142 | ||
NM_001313414.3 | NP_001300343.1 | 10 - 11 | 1898 | 142 | ||
NM_001313415.4 | NP_001300344.1 | 8 - 9 | 1624 | 142 | ||
NM_001313417.4 | NP_001300346.1 | 2 - 3 | 124 | 142 | ||
NM_001380898.1 | NP_001368578.1 | 8 - 9 | 1615 | 142 | ||
NM_075760.6 | NP_508161.3 | 11 - 12 | 2000 | 142 | ||
GenBank mRNA | AF005205.1 | - | 8 - 9 | 1663 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |