Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sms-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075781.5 | NP_508182.1 | 5 - 6 | 881 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |