Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncs-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075785.4 | NP_508186.1 | 3 - 4 | 300 | 124 | |
GenBank mRNA | L33680.1 | - | 3 - 4 | 231 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |