Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncs-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075785.4 | NP_508186.1 | 5 - 6 | 544 | 90 | |
GenBank mRNA | L33680.1 | - | 5 - 6 | 475 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Caenorhabditis elegans Probe spans exons |