Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rme-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383525.1 | 7 - 8 | 1128 | 91 | ||
NM_075807.5 | NP_508208.2 | 7 - 8 | 1122 | 91 | ||
GenBank mRNA | AY750054.1 | - | 7 - 8 | 1122 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |