Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lmp-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380905.1 | NP_001367433.1 | 5 - 6 | 696 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |