Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029571.4 | NP_001024742.1 | 8 - 9 | 1332 | 66 | |
NM_001029572.4 | NP_001024743.1 | 8 - 9 | 1353 | 66 | ||
GenBank mRNA | AF130406.1 | - | 8 - 9 | 1344 | 66 | |
AF130407.1 | - | 8 - 9 | 1332 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |