Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdf-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075996.3 | NP_508397.2 | 2 - 3 | 170 | 69 | |
GenBank mRNA | EF141321.1 | - | 2 - 3 | 170 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |