Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glr-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001350348.3 | NP_001337310.1 | 12 - 13 | 1922 | 104 | |
NM_001373243.1 | NP_001359931.1 | 8 - 9 | 1373 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |