Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fox-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330894.3 | NP_001317857.1 | 1 | 328 | 92 | |
NM_001330895.5 | NP_001317738.1 | 2702 | 92 | |||
NM_001330896.3 | NP_001317861.1 | 1 - 2 | 61 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons |