Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hid-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029577.3 | NP_001024748.1 | 3 - 4 | 484 | 105 | |
NM_001029578.5 | NP_001024749.1 | 3 - 4 | 438 | 105 | ||
GenBank mRNA | AY070228.1 | - | 3 - 4 | 454 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |